Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QYILGEWDFQ GLSLRMVPPV FIPRPETEEL VEWVLEEVAQ RSHAVGSPGS PLILEVGCGS GAISLSLLSQ LPQSRVIAVD KREAAISLTH E
Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 81%.
N5-glutamine methyltransferase responsible for the methylation of the glutamine residue in the universally conserved GGQ motif of the mitochondrial translation release factor MTRF1L. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HEMK homolog 7kb; HemK methyltransferase family member 1; M.HsaHemKP; MTRF1L release factor glutamine methyltransferase; testis secretory sperm-binding protein Li 225n
Gene Aliases: HEMK; HEMK1; MTQ1
UniProt ID: (Human) Q9Y5R4
Entrez Gene ID: (Human) 51409
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support