Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The helix-loop-helix (HLH) structures are known motifs commonly found in membrane-active and DNA-binding proteins. Helix-loop-helix proteins (HEN1 and HEN2) are DNA-binding proteins that may be involved in cell-type determination in the early nervous system. Studies of expression in normal tissues have demonstrated expression of NSCL-1 and NSCL-2 (genes encoding HEN1 and HEN2) in the developing central and peripheral nervous system, specifically in developing neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa34; bHLHa35; Class A basic helix-loop-helix protein 34; Class A basic helix-loop-helix protein 35; Helix-loop-helix protein 1; HELIX-LOOP-HELIX PROTEIN 1 (HEN1) (NSCL); Helix-loop-helix protein 2; HEN-1; HEN-2; Nescient helix loop helix 1; Nescient helix loop helix 2; NSCL-1; NSCL-2
Gene Aliases: 6230401I09Rik; BHLHA34; BHLHA35; HEN1; HEN2; KIAA0490; NHLH1; NHLH2; NSCL; Nscl-2; NSCL1; NSCL2; Tal2
UniProt ID: (Human) Q02575, (Human) Q02577, (Mouse) Q02576, (Mouse) Q64221
Entrez Gene ID: (Human) 4807, (Human) 4808, (Rat) 289230, (Mouse) 18071, (Mouse) 18072, (Rat) 295327
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support