Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MESETEPEPV TLLVKSPNQR HRDLELSGDR GWSVGHLKAH LSRVYPERPR PEDQRLIYSG KLLLDHQCLR DLLPKQEKRH VLHLVCNVKS PSKMPEINAK VAESTEEPAG SNRGQYPEDS SSDGLRQREV LRNLSSPGWE NISRPEAAQQ AFQGLGPGFS GYTPYGWLQL SWFQQIYARQ YYMQYLAATA ASGAFVPPPS AQEIPVVSAP APAPIHNQFP AENQPANQNA APQVVVNPGA NQNLRMNAQG GPIVEEDDEI NRDWLDWTYS AATFSVFLSI LYFYSSLSRF LMVMGATVVM YLHHVGWFPF RPRPVQNFPN DGPPPDVVNQ DPNNNLQEGT DPETEDPNHL PPDRDVLDGE QTSPSFMSTA WLVFKTFFAS LLPEGPPAIA N
The accumulation of unfolded proteins in the endoplasmic reticulum (ER) triggers the ER stress response. This response includes the inhibition of translation to prevent further accumulation of unfolded proteins, the increased expression of proteins involved in polypeptide folding, known as the unfolded protein response (UPR), and the destruction of misfolded proteins by the ER-associated protein degradation (ERAD) system. This gene may play a role in both UPR and ERAD. Its expression is induced by UPR and it has an ER stress response element in its promoter region while the encoded protein has an N-terminal ubiquitin-like domain which may interact with the ERAD system. This protein has been shown to interact with presenilin proteins and to increase the level of amyloid-beta protein following its overexpression. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms. The full-length nature of all transcript variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homocysteine-inducible endoplasmic reticulum stress-inducible ubiquitin-like domain member 1 protein; homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1; Homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domain member 1 protein; Methyl methanesulfonate (MMF)-inducible fragment protein 1; MMS-inducible
Gene Aliases: HERP; HERPUD1; KIAA0025; MIF1; SUP
UniProt ID: (Human) Q15011
Entrez Gene ID: (Human) 9709
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support