Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RRDRINSSIE QLKLLLEQEF ARHQPNSKLE KADILEMAVS YLKHSKAFVA AAGPKSLHQD YSEGYSWCLQ EAVQFLTLHA ASDTQMKLLY HFQR
Hairy and Enhancer-of-split homologues 5 (HES5) is a helix-loop-helix transcriptional factor that negatively regulates cell differentiation during embryogenesis. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. HES5 is reported to be a down stream effector of the Notch Signaling Pathway and is expressed in advanced ovarian serous adenocarcinomas as well as in cervical squamous carcinoma. Reports also suggest that HES5 is abundantly expressed in osteoarthritis along with Notch1 and Jagged1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHb38; Class B basic helix-loop-helix protein 38; Hairy and enhancer of split 5; RP3-395M20.5; Transcription factor HES-5
Gene Aliases: BHLHB38; HES5
UniProt ID: (Human) Q5TA89
Entrez Gene ID: (Human) 388585
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support