Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MELCGLGLP RPPMLLALLL ATLLAAMLAL LTQVALVVQV AEAARAPSVS AKPGPALWPL PLSVKMTPNL LHLAPENFYI SHSPNSTAGP SCTLLEEAFR RYHGYIFGFY KWHHEPAEFQ AKTQVQQLLV SITLQSECDA FPNISSDESY TLLVKEPVAV LKANRVWGAL RGLETFSQLV YQDSYGTFTI NESTIIDSPR FSHRGILIDT SRHYLPVKII LKTLDAMAFN KFNVLHWHIV DDQSFPYQSI TFPELSNKGS YSLSHVYTPN DVRMVIEYAR LRGIRVLPEF DTPGHTLSWG KGQKDLLTPC YSRQNKLDSF GPINPTLNTT YSF (1-332 aa encoded by BC017378)
Hexosaminidase B (HEXB), also designated beta-hexosaminidase B, is a Hexosaminidase B (HEXB), also designated b-hexosaminidase B, is a tetramer of two b-A and two b-B chains and is found in the lysosomes of cells. Sandhoff disease (SD), also known as GM2-gangliosidosis type II, is caused by mutations in the HEXB gene that affect the b subunit. These mutations disrupt the activity of HEXB and HEXA, which prevents the breakdown of GM2 ganglioside, a fatty material found in the brain, therby rendering both the HEXA and HEXB enzymes deficient. SD is a rare autosomal recessive disorder characterized by an accumulation of GM2 ganglioside, which causes progressive destruction of the central nervous system. Sandhoff disease is similar to Tay-Sachs disease, which is caused by mutations in the HEXA gene, although SD is more severe.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-hexosaminidase subunit beta; Beta-N-acetylhexosaminidase subunit beta; Cervical cancer proto-oncogene 7 protein; epididymis luminal protein 248; epididymis secretory protein Li 111; HCC-7; hexosaminidase B (beta polypeptide); Hexosaminidase subunit B; N-acetyl-beta-glucosaminidase subunit beta
Gene Aliases: ENC-1AS; HCC7; HEL-248; HEL-S-111; HEXB
UniProt ID: (Human) P07686, (Mouse) P20060, (Rat) Q6AXR4
Entrez Gene ID: (Human) 3074, (Mouse) 15212, (Rat) 294673
Molecular Function:
glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support