Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
HEXD (Hexosaminidase D) is a Protein Coding gene. Among its related pathways are Other glycan degradation and Metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-hexosaminidase D; Beta-N-acetylhexosaminidase; hexosaminidase (glycosyl hydrolase family 20, catalytic domain) containing; Hexosaminidase D; hexosaminidase D, cytosolic; Hexosaminidase domain-containing protein; N-acetyl-beta-galactosaminidase
Gene Aliases: BC069960; HEXD; HEXDC
UniProt ID: (Human) Q8WVB3, (Mouse) Q3U4H6
Entrez Gene ID: (Human) 284004, (Mouse) 238023, (Rat) 100216475
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support