Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PCEETTSESDM DETIDVGSEN NYSGQSTSSV IRLNSPTTTS QIMARKK
Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
Downstream effector of Notch signaling which may be required for cardiovascular development. Transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. Represses transcription by the cardiac transcriptional activators GATA4 and GATA6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHb32; cardiovascular basic helix-loop-helix factor 1; Cardiovascular helix-loop-helix factor 1; Class B basic helix-loop-helix protein 32; h hHRT2; Hairy and enhancer of split-related protein 2; Hairy-related transcription factor 2; hairy/enhancer-of-split related with YRPW motif 2; Hairy/enhancer-of-split related with YRPW motif protein 2; hCHF1; hes-related family bHLH transcription factor with YRPW motif 2; HES-related repressor protein 1; HES-related repressor protein 2; HESR-2; hHRT2; HRT-2; Protein gridlock homolog
Gene Aliases: BHLHB32; CHF1; GRIDLOCK; GRL; HERP; HERP1; HESR2; HEY2; HRT2
UniProt ID: (Human) Q9UBP5
Entrez Gene ID: (Human) 23493
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support