Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31758 has been tested in indirect ELISA.
Histones are relatively small proteins that contain a high proportion of positively charged amino acids, e. g., arginine and lysine. This positive charge allows histones to bind tightly to the negatively charged DNA. There are five basic types of histone molecules called H1, H2A, H2B, H3, and H4. Histones are abundantly present in virtually all eukaryotic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H3 histone family, member T; H3/g; H3/t; histone 3, H3; Histone H3.1t; Histone H3.4
Gene Aliases: H3-4; H3.4; H3/g; H3FT; H3t; HIST3H3
UniProt ID: (Human) Q16695
Entrez Gene ID: (Human) 8290
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support