Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MQYPHPGPAA GAVGVPLYAP TPLLQPAHPT PFYIEDILGR GPAAPTPAPT LPSPNSSFTS LVSPYRTPVY EPTPIHPAFS HHSAAALAAA YGPGGFGGPL YPFPRTVNDY THALLRHDPL GKPLLWSPFL QRPLHKRKGG QVRFSNDQTI ELEKKFETQK YLSPPERKRL TKMLQLSERQ VKTWFQNRRA KWRRLKQENP QSNKKEELES LDSSCDQRQD LPSEQNKGAS LDSSQCSPSP ASQEDLESEI SEDSDQEVDI EGDKSYFNAG
HHEX is a member of the homeobox family of transcription factors, many of which are involved in developmental processes. Expression in specific hematopoietic lineages suggests that this protein may play a role in hematopoietic differentiation. Establishes anterior identity at two levels 1) acts early to enhance canonical WNT-signaling by repressing expression of TLE4 and 2) acts later to inhibit NODAL-signaling by directly targeting NODAL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hematopoietically-expressed homeobox protein HHEX; homeobox; Homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; Proline-rich homeodomain protein; proline-rich homeodomain-containing transcription factor
Gene Aliases: HEX; HHEX; HMPH; HOX11L-PEN; PRH; PRHX
UniProt ID: (Human) Q03014
Entrez Gene ID: (Human) 3087
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support