Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAASLRLLGA ASGLRYWSRR LRPAAGSFAA VCSRSVASKT PVGFIGLGNM GNPMAKNLMK HGYPLIIYDV FPDACKEFQD AGEQVVSSPA DVAEKADRII TMLPTSINAI EAYSGANGIL KKVKKGSLLI DSSTIDPAVS KELAKEVEKM GAVFMDAPVS GGVGAARSGN LTFMVGGVED EFAAAQELLG CMGSNVVYCG AVGTGQAAKI CNNMLLAISM IGTAEAMNLG IRLGLDPKLL AKILNMSSGR CWSSDTYNPV PGVMDGVPSA NNYQGGFGTT LMAKDLGLAQ DSATSTKSPI LLGSLAHQIY RMMCAKGYSK KDFSSVFQFL REEETF
3-hydroxyisobutyrate dehydrogenase (3-hydroxy-2-methylpropanoate:NAD(+) oxidoreductase, EC 1.1. 1.31) is a dimeric mitochondrial enzyme that catalyzes the NAD(+)-dependent, reversible oxidation of 3-hydroxyisobutyrate, an intermediate of valine catabolism, to methylmalonate semialdehyde.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-hydroxyisobutyrate dehydrogenase; 3-hydroxyisobutyrate dehydrogenase, mitochondrial; HIBADH
Gene Aliases: HIBADH; NS5ATP1
UniProt ID: (Human) P31937
Entrez Gene ID: (Human) 11112
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support