Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSTDTGVSLP SYEEDQGSKL IRKAKEAPFV PVGIAGFAAI VAYGLYKLKS RGNTKMSIHL IHMRVAAQGF VVGAMTVGMG YSMYREFWAK PKP
Proposed subunit of cytochrome c oxidase (COX, complex IV), which is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. May play a role in the assembly of respiratory supercomplexes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP564K247; HIG1 domain family member 1A, mitochondrial; HIG1 domain family, member 1A; HIG1 hypoxia inducible domain family, member 1A; hypoxia inducible gene 1; Hypoxia-inducible gene 1 protein; RCF1 homolog A; RCF1a
Gene Aliases: HIG1; HIGD1A; HSPC010; RCF1a
UniProt ID: (Human) Q9Y241
Entrez Gene ID: (Human) 25994
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support