Product References
The regulation of cell homeostasis and antiviral innate immunity by autophagy during classical swine fever virus infection.
Emerging microbes & infections
Li X,Song Y,Wang X,Fu C,Zhao F,Zou L,Wu K,Chen W,Li Z,Fan J,Li Y,Li B,Zeng S,Liu X,Zhao M,Yi L,Chen J,Fan S
Fri Dec 01 00:00:00 EST 2023
Rare sugar L-sorbose exerts antitumor activity by impairing glucose metabolism.
Communications biology
Xu HL,Zhou X,Chen S,Xu S,Li Z,Nakanishi H,Gao XD
Sat Mar 11 00:00:00 EST 2023
Ectopic endometriotic stromal cells-derived lactate induces M2 macrophage polarization via Mettl3/Trib1/ERK/STAT3 signalling pathway in endometriosis.
Immunology
Gou Y,Wang H,Wang T,Wang H,Wang B,Jiao N,Yu Y,Cao Y,Wang H,Zhang Z
Wed Mar 01 00:00:00 EST 2023
HIF-1α Mediates Arsenic-Induced Metabolic Reprogramming in Lung Bronchial Epithelial Cells.
Biological trace element research
Wang W,Zhang A
Mon May 01 00:00:00 EDT 2023
NR3C2 inhibits the proliferation of colorectal cancer via regulating glucose metabolism and phosphorylating AMPK.
Journal of cellular and molecular medicine
Liu H,Lei W,Li Z,Wang X,Zhou L
Sat Apr 01 00:00:00 EDT 2023
Alamandine/MrgD axis prevents TGF-β1-mediated fibroblast activation via regulation of aerobic glycolysis and mitophagy.
Journal of translational medicine
Wang W,Zhang Y,Huang W,Yuan Y,Hong Q,Xie Z,Li L,Chen Y,Li X,Meng Y
Fri Jan 13 00:00:00 EST 2023
Alamandine/MrgD axis prevents TGF-β1-mediated fibroblast activation via regulation of aerobic glycolysis and mitophagy.
Journal of translational medicine
Wang W,Zhang Y,Huang W,Yuan Y,Hong Q,Xie Z,Li L,Chen Y,Li X,Meng Y
Fri Jan 13 00:00:00 EST 2023
Alamandine/MrgD axis prevents TGF-β1-mediated fibroblast activation via regulation of aerobic glycolysis and mitophagy.
Journal of translational medicine
Wang W,Zhang Y,Huang W,Yuan Y,Hong Q,Xie Z,Li L,Chen Y,Li X,Meng Y
Fri Jan 13 00:00:00 EST 2023
Alamandine/MrgD axis prevents TGF-β1-mediated fibroblast activation via regulation of aerobic glycolysis and mitophagy.
Journal of translational medicine
Wang W,Zhang Y,Huang W,Yuan Y,Hong Q,Xie Z,Li L,Chen Y,Li X,Meng Y
Fri Jan 13 00:00:00 EST 2023
TMT-based quantitative proteomic analysis of spheroid cells of endometrial cancer possessing cancer stem cell properties.
Stem cell research & therapy
Cao M,Liu Z,You D,Pan Y,Zhang Q
Thu May 04 00:00:00 EDT 2023
Bleomycin restricts the glycolysis of lymphatic endothelial cells by inhibiting dimeric PKM2 formation: A novel mechanism for lymphatic malformation treatment.
Biochemical pharmacology
Chai M,Wang R,Jiang H,Zhu T,Liu J,Huang C,Zhong W,Cai Y
Sat Oct 01 00:00:00 EDT 2022
Phosphorylation and acetylation of glycolytic enzymes cooperatively regulate their activity and lamb meat quality.
Food chemistry
Ren C,Li X,Bai Y,Schroyen M,Zhang D
Thu Dec 15 00:00:00 EST 2022
Novel long non-coding RNA CYB561-5 promotes aerobic glycolysis and tumorigenesis by interacting with basigin in non-small cell lung cancer.
Journal of cellular and molecular medicine
Li L,Li Z,Qu J,Wei X,Suo F,Xu J,Liu X,Chen C,Zheng S
Tue Mar 01 00:00:00 EST 2022
Engineering of thick human functional myocardium via static stretching and electrical stimulation.
iScience
Pretorius D,Kahn-Krell AM,LaBarge WC,Lou X,Zhang J
Fri Mar 18 00:00:00 EDT 2022
Lactate Dehydrogenase B Is Required for Pancreatic Cancer Cell Immortalization Through Activation of Telomerase Activity.
Frontiers in oncology
Wang R,Li J,Zhang C,Guan X,Qin B,Jin R,Qin L,Xu S,Zhang X,Liu R,Ye Q,Cheng L
Sat Jul 16 00:00:00 EDT 2022
Tanshinone IIA inhibits cell growth by suppressing SIX1-induced aerobic glycolysis in non-small cell lung cancer cells.
Oncology letters
Qi H,Chen Z,Qin Y,Wang X,Zhang Z,Li Y
Wed Jun 01 00:00:00 EDT 2022
Bmal1 inhibits phenotypic transformation of hepatic stellate cells in liver fibrosis via IDH1/α-KG-mediated glycolysis.
Acta pharmacologica Sinica
Xu L,Yang TY,Zhou YW,Wu MF,Shen J,Cheng JL,Liu QX,Cao SY,Wang JQ,Zhang L
Tue Feb 01 00:00:00 EST 2022
The MBNL1/circNTRK2/PAX5 pathway regulates aerobic glycolysis in glioblastoma cells by encoding a novel protein NTRK2-243aa.
Cell death & disease
Zhao Y,Song J,Dong W,Liu X,Yang C,Wang D,Xue Y,Ruan X,Liu L,Wang P,Zhang M,Liu Y
Mon Sep 05 00:00:00 EDT 2022
Myosin 1b promotes migration, invasion and glycolysis in cervical cancer via ERK/HIF-1α pathway.
American journal of translational research
Wen LJ,Hu XL,Li CY,Liu J,Li ZY,Li YZ,Zhou JY
Tue Dec 28 00:00:00 EST 2021
RSL3 enhances the antitumor effect of cisplatin on prostate cancer cells via causing glycolysis dysfunction.
Biochemical pharmacology
Li M,Chen X,Wang X,Wei X,Wang D,Liu X,Xu L,Batu W,Li Y,Guo B,Zhang L
Fri Oct 01 00:00:00 EDT 2021
Study on the Antitumor Effect and Glycolysis of Andrographolide in Anaplastic Thyroid Carcinoma.
Evidence-based complementary and alternative medicine : eCAM
Yang K,Wu K,Feng J,Yutian L,Zhu X,Xu D
Mon Apr 25 00:00:00 EDT 2022
Bmal1 Regulates Macrophage Polarize Through Glycolytic Pathway in Alcoholic Liver Disease.
Frontiers in pharmacology
Zhou Y,Wu M,Xu L,Cheng J,Shen J,Yang T,Zhang L
Fri Apr 02 00:00:00 EDT 2021
Integrated Metabolomics and Transcriptomics Analyses Reveal Metabolic Landscape in Neuronal Cells during JEV Infection.
Virologica Sinica
Li M,Yang J,Ye C,Bian P,Yang X,Zhang H,Luo C,Xue Z,Lei Y,Lian J
Wed Dec 01 00:00:00 EST 2021
PGC1α Promotes Cisplatin Resistance in Ovarian Cancer by Regulating the HSP70/HK2/VDAC1 Signaling Pathway.
International journal of molecular sciences
Li Y,Kang J,Fu J,Luo H,Liu Y,Li Y,Sun L
Wed Mar 03 00:00:00 EST 2021
MiR-489-3p Reduced Pancreatic Cancer Proliferation and Metastasis By Targeting PKM2 and LDHA Involving Glycolysis.
Frontiers in oncology
Zhang D,He Z,Shen Y,Wang J,Liu T,Jiang J
Thu Apr 28 00:00:00 EDT 2022
Activated Pancreatic Stellate Cells Enhance the Warburg Effect to Cause the Malignant Development in Chronic Pancreatitis.
Frontiers in oncology
Tao Y,Shao F,Cai M,Liu Z,Peng Y,Huang Q,Meng F
Tue Sep 21 00:00:00 EDT 2021
Activated Pancreatic Stellate Cells Enhance the Warburg Effect to Cause the Malignant Development in Chronic Pancreatitis.
Frontiers in oncology
Tao Y,Shao F,Cai M,Liu Z,Peng Y,Huang Q,Meng F
Tue Sep 21 00:00:00 EDT 2021
Activated Pancreatic Stellate Cells Enhance the Warburg Effect to Cause the Malignant Development in Chronic Pancreatitis.
Frontiers in oncology
Tao Y,Shao F,Cai M,Liu Z,Peng Y,Huang Q,Meng F
Tue Sep 21 00:00:00 EDT 2021
Activated Pancreatic Stellate Cells Enhance the Warburg Effect to Cause the Malignant Development in Chronic Pancreatitis.
Frontiers in oncology
Tao Y,Shao F,Cai M,Liu Z,Peng Y,Huang Q,Meng F
Tue Sep 21 00:00:00 EDT 2021
Ginsenoside Rh4 suppresses aerobic glycolysis and the expression of PD-L1 via targeting AKT in esophageal cancer.
Biochemical pharmacology
Deng X,Zhao J,Qu L,Duan Z,Fu R,Zhu C,Fan D
Sat Aug 01 00:00:00 EDT 2020
Ginsenoside Rh4 suppresses aerobic glycolysis and the expression of PD-L1 via targeting AKT in esophageal cancer.
Biochemical pharmacology
Deng X,Zhao J,Qu L,Duan Z,Fu R,Zhu C,Fan D
Sat Aug 01 00:00:00 EDT 2020