Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: This SK10 monoclonal antibody reacts with the human MHC Class II HLA-DQ molecule. This cell surface receptor is a heterodimeric protein composed of an alpha (HLA-DQA1) and beta (HLA-DQB1) subunit. HLA-DQ is expressed on antigen presenting cells such as B cells, monocytes, macrophages, dendritic cells, and activated T cells. HLA-DQ plays a role in presenting peptide antigens to CD4+ T lymphocytes. Reports indicate that certain alleles of HLA-DQ are associated with celiac disease and type I diabetes.
Applications Reported: This SK10 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This SK10 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
Filtration: 0.2 µm post-manufacturing filtered.
HLA-DQ is one of the HLA (human leukocyte antigen) molecules belonging to class II of the major histocompatibility protein (MHC) surface proteins. HLA-DQ and the other MHC class II molecules are heterodimeric proteins consisting of an alpha (DQA) and a beta chain (DQB), both anchored in the membrane. They play a central role in the immune system by presenting peptides derived from extracellular proteins. MHC Class II molecules are expressed in antigen-presenting cells (APC: B lymphocytes, dendritic cells, macrophages) and present peptides endocytosed, digested, and loaded onto the proteins prior to their migration to the cell surface. Other human MHC class II molecules are HLA-DP, HLA-DM, HLA-DR, HLA-DOA, and HLA-DOB. HLA-DQ displays significant variability, particularly in the beta subunit within the region that affects the peptides bound and presented by the protein. Different classes of DQ based on similar beta subunits allow for the grouping of these variants into serotypes, which can often be recognized by antibodies raised against these subunits.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HLA class II antigen; HLA class II histocompatibility antigen, DQ beta 1 chain; lymphocyte antigen; MHC class II antigen DQB1; MHC class II antigen HLA-DQ-beta-1; MHC class II DQ beta chain; MHC class II HLA-DQ beta glycoprotein; MHC class2 antigen; MHC DQ beta
Gene Aliases: CELIAC1; HLA-DQB; HLA-DQB1; HLA-DRB1; IDDM1
UniProt ID: (Human) Q08GC8
Entrez Gene ID: (Human) 3119
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support