Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
HMGB3 belongs to the high mobility group protein superfamily. Like HMG1 and HMG2, HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High mobility group protein 2a; High mobility group protein 4; High mobility group protein B3; high mobilty group protein 2A; high-mobility group (nonhistone chromosomal) protein 4; high-mobility group protein 4; HMG-2a; HMG-4; MGC90319
Gene Aliases: HMG-2a; HMG-4; HMG2A; HMG4; HMGB3
UniProt ID: (Human) O15347, (Mouse) O54879
Entrez Gene ID: (Human) 3149, (Mouse) 15354
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support