Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAAMRKALPR RLVGLASLRA VSTSSMGTLP KRVKIVEVGP RDGLQNEKNI VSTPVKIKLI DMLSEAGLSV IETTSFVSPK WVPQMGDHTE VLKGIQKFPG INYPVLTPNL KGFEAAVAAG AKEVVIFGAA SELFTKKNIN CSIEESFQRF DAILKAAQSA NISVRGYVSC ALGCPYEGKI SPAKVAEVTK KFYSMGCYEI SLGDTIGVGT PGIMKDMLSA VMQEVPLAAL AVHCHDTYGQ ALANTLMALQ MGVSVVDSSV AGLGGCPYAQ GASGNLATED LVYMLEGLGI HTGVNLQKLL EAGNFICQAL NRKTSSKVAQ ATCKL
The protein encoded by this gene belongs to the HMG-CoA lyase family. It is a mitochondrial enzyme that catalyzes the final step of leucine degradation and plays a key role in ketone body formation. Mutations in this gene are associated with HMG-CoA lyase deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-hydroxy-3-methylglutarate-CoA lyase; 3-hydroxy-3-methylglutaryl-CoA lyase; 3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase; HL; HMG-CoA lyase; hydroxymethylglutaricaciduria; Hydroxymethylglutaryl-CoA lyase, mitochondrial; mitochondrial 3-hydroxy-3-methylglutaryl-CoA lyase
Gene Aliases: HL; HMGCL
UniProt ID: (Human) P35914
Entrez Gene ID: (Human) 3155
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support