Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Keep as concentrated solution.
Predicted reactivity: Mouse (81%), Rat (80%), Dog (85%), Pig (82%), Bovine (83%).
Positive Control: HeLa, HeLa 500 uM CoCl2 treatment for 24 hr.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heat shock protein, 32-kD; heme orygenase 1; heme oxygenase (decycling) 1; Heme oxygenase 1; HO-1
Gene Aliases: bK286B10; HMOX1; HMOX1D; HO; HO-1; HO1; HSP32
UniProt ID: (Human) P09601
Entrez Gene ID: (Human) 3162
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support