Product References
LncRNA BC promotes lung adenocarcinoma progression by modulating IMPAD1 alternative splicing.
Clinical and translational medicine
Chen QW,Cai QQ,Yang Y,Dong S,Liu YY,Chen ZY,Kang CL,Qi B,Dong YW,Wu W,Zhuang LP,Shen YH,Meng ZQ,Wu XZ
Sun Jan 01 00:00:00 EST 2023
Characterization of the protein expression and localization of hnRNP family members during murine spermatogenesis.
Asian journal of andrology
Wang XL,Li JM,Yuan SQ
Fri Apr 28 00:00:00 EDT 2023
Characterization of the protein expression and localization of hnRNP family members during murine spermatogenesis.
Asian journal of andrology
Wang XL,Li JM,Yuan SQ
Fri Apr 28 00:00:00 EDT 2023
Alternative splicing liberates a cryptic cytoplasmic isoform of mitochondrial MECR that antagonizes influenza virus.
PLoS biology
Baker SF,Meistermann H,Tzouros M,Baker A,Golling S,Polster JS,Ledwith MP,Gitter A,Augustin A,Javanbakht H,Mehle A
Thu Dec 01 00:00:00 EST 2022
Alternative splicing liberates a cryptic cytoplasmic isoform of mitochondrial MECR that antagonizes influenza virus.
PLoS biology
Baker SF,Meistermann H,Tzouros M,Baker A,Golling S,Polster JS,Ledwith MP,Gitter A,Augustin A,Javanbakht H,Mehle A
Thu Dec 01 00:00:00 EST 2022
RNA-binding protein hnRNP UL1 binds κB sites to attenuate NF-κB-mediated inflammation.
Journal of autoimmunity
Ma Z,Zhou Y,Wang Y,Xu Y,Liu Y,Liu Y,Jiang M,Zhang X,Cao X
Sun May 01 00:00:00 EDT 2022
RNA-binding protein hnRNP UL1 binds κB sites to attenuate NF-κB-mediated inflammation.
Journal of autoimmunity
Ma Z,Zhou Y,Wang Y,Xu Y,Liu Y,Liu Y,Jiang M,Zhang X,Cao X
Sun May 01 00:00:00 EDT 2022
HNRNPUL1 inhibits cisplatin sensitivity of esophageal squamous cell carcinoma through regulating the formation of circMAN1A2.
Experimental cell research
Li J,Sang M,Zheng Y,Meng L,Gu L,Li Z,Liu F,Wu Y,Li W,Shan B
Wed Dec 01 00:00:00 EST 2021
Arginine methylation of hnRNPUL1 regulates interaction with NBS1 and recruitment to sites of DNA damage.
Scientific reports
Gurunathan G,Yu Z,Coulombe Y,Masson JY,Richard S
Thu May 28 00:00:00 EDT 2015
Cellular RNA binding proteins NS1-BP and hnRNP K regulate influenza A virus RNA splicing.
PLoS pathogens
Tsai PL,Chiou NT,Kuss S,García-Sastre A,Lynch KW,Fontoura BM
Fri Jan 17 00:00:00 EST 2014
Stability of a long noncoding viral RNA depends on a 9-nt core element at the RNA 5' end to interact with viral ORF57 and cellular PABPC1.
International journal of biological sciences
Massimelli MJ,Kang JG,Majerciak V,Le SY,Liewehr DJ,Steinberg SM,Zheng ZM
Tue Feb 21 00:00:00 EST 2012
Stability of a long noncoding viral RNA depends on a 9-nt core element at the RNA 5' end to interact with viral ORF57 and cellular PABPC1.
International journal of biological sciences
Massimelli MJ,Kang JG,Majerciak V,Le SY,Liewehr DJ,Steinberg SM,Zheng ZM
Tue Feb 21 00:00:00 EST 2012
Vesicular stomatitis virus inhibits mitotic progression and triggers cell death.
EMBO reports
Chakraborty P,Seemann J,Mishra RK,Wei JH,Weil L,Nussenzveig DR,Heiber J,Barber GN,Dasso M,Fontoura BM
Thu Oct 01 00:00:00 EDT 2009
Vesicular stomatitis virus inhibits mitotic progression and triggers cell death.
EMBO reports
Chakraborty P,Seemann J,Mishra RK,Wei JH,Weil L,Nussenzveig DR,Heiber J,Barber GN,Dasso M,Fontoura BM
Thu Oct 01 00:00:00 EDT 2009
Vesicular stomatitis virus inhibits mitotic progression and triggers cell death.
EMBO reports
Chakraborty P,Seemann J,Mishra RK,Wei JH,Weil L,Nussenzveig DR,Heiber J,Barber GN,Dasso M,Fontoura BM
Thu Oct 01 00:00:00 EDT 2009
Vesicular stomatitis virus inhibits mitotic progression and triggers cell death.
EMBO reports
Chakraborty P,Seemann J,Mishra RK,Wei JH,Weil L,Nussenzveig DR,Heiber J,Barber GN,Dasso M,Fontoura BM
Thu Oct 01 00:00:00 EDT 2009
Influenza virus targets the mRNA export machinery and the nuclear pore complex.
Proceedings of the National Academy of Sciences of the United States of America
Satterly N,Tsai PL,van Deursen J,Nussenzveig DR,Wang Y,Faria PA,Levay A,Levy DE,Fontoura BM
Tue Feb 06 00:00:00 EST 2007
Influenza virus targets the mRNA export machinery and the nuclear pore complex.
Proceedings of the National Academy of Sciences of the United States of America
Satterly N,Tsai PL,van Deursen J,Nussenzveig DR,Wang Y,Faria PA,Levay A,Levy DE,Fontoura BM
Tue Feb 06 00:00:00 EST 2007
Influenza virus targets the mRNA export machinery and the nuclear pore complex.
Proceedings of the National Academy of Sciences of the United States of America
Satterly N,Tsai PL,van Deursen J,Nussenzveig DR,Wang Y,Faria PA,Levay A,Levy DE,Fontoura BM
Tue Feb 06 00:00:00 EST 2007
Influenza virus targets the mRNA export machinery and the nuclear pore complex.
Proceedings of the National Academy of Sciences of the United States of America
Satterly N,Tsai PL,van Deursen J,Nussenzveig DR,Wang Y,Faria PA,Levay A,Levy DE,Fontoura BM
Tue Feb 06 00:00:00 EST 2007
Influenza virus targets the mRNA export machinery and the nuclear pore complex.
Proceedings of the National Academy of Sciences of the United States of America
Satterly N,Tsai PL,van Deursen J,Nussenzveig DR,Wang Y,Faria PA,Levay A,Levy DE,Fontoura BM
Tue Feb 06 00:00:00 EST 2007
Influenza virus targets the mRNA export machinery and the nuclear pore complex.
Proceedings of the National Academy of Sciences of the United States of America
Satterly N,Tsai PL,van Deursen J,Nussenzveig DR,Wang Y,Faria PA,Levay A,Levy DE,Fontoura BM
Tue Feb 06 00:00:00 EST 2007