Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGGRKRERKA AVEEDTSLSE SEGPRQPDGD EEESTALSIN EEMQRMLNQL REYDFEDDCD SLTWEETEET LLLWEDFSGY AMAAAEAQGE QQEDSLEKVI KDTESLFKTR EKEYQETIDQ IELELATAKN DMNRHLHEYM EMCSMKRGLD VQMETCRRLI TQSGDRKSPA FTAVPLSDPP PPPSEAEDSD RDVSSDSSMR
This gene is a member of the intermediate filament family. Intermediate filaments are proteins which are primordial components of the cytoskeleton and nuclear envelope. The proteins encoded by the members of this gene family are evolutionarily and structurally related but have limited sequence homology, with the exception of the central rod domain. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Intermediate filament family orphan 1; intermediate filament-like MGC:2625; NHEJ factor IFFO1; Non-homologous end joining factor IFFO1; Tumor antigen HOM-TES-103
Gene Aliases: HOM-TES-103; IFFO; IFFO1
UniProt ID: (Human) Q0D2I5
Entrez Gene ID: (Human) 25900
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support