Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MFSVESLERA ELCESLLTWI QTFNVDAPCQ TVEDLTNGVV MAQVLQKIDP AYFDENWLNR IKTEVGDNWR LKISNLKKIL KGILDYNHEI LGQQINDFTL PDVNLIGEHS DAAELGRMLQ LILGCAVNCE QKQEYIQAIM MMEESVQHVV MTAIQELMSK ESPVSAGNDA YVDLDRQLKK TTEELNEALS AKEEIAQRCH ELDMQVAALQ EEKSSLLAEN QVLMERLNQS DSIEDPNSPA GRRHLQLQTQ LEQLQEETFR LEAAKDDYRI RCEELEKEIS ELRQQNDELT TLADEAQSLK DEIDVLRHSS DKVSKLEGQV ESYKKKLEDL GDLRRQVKLL EEKNTMYMQN TVSLEEELRK ANAARSQLET YKRQVVELQN RLSEESKKAD KLDFEYKRLK EKVDSLQKEK DRLRTERDSL KETIEELRCV QAQEGQLTTQ GLMPLGSQES SDSLAAEIVT PEIREKLIRL QHENKMLKLN QEGSDNEKIA LLQSLLDDAN LRKNELETEN RLVNQRLLEV QSQVEELQKS LQDQGSKAED SVLLKKKLEE HLEKLHEANN ELQKKRAIIE DLEPRFNNSS LKIEELQEAL RKKEEEMKQM EERYKKYLEK AKSVIRTLDP KQNQGAAPEI QALKNQLQER DRLFHSLEKE YEKTKSQREM EEKYIVSAWY NMGMTLHKKA AEDRLASTGS GQSFLARQRQ ATSSRRSYPG HVQPATAR
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: h-hook3; hHK3; hook homolog 3; Protein Hook homolog 3
Gene Aliases: HK3; HOOK3
UniProt ID: (Human) Q86VS8
Entrez Gene ID: (Human) 84376
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support