Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IALLQSLLDD ANLRKNELET ENRLVNQRLL EVQSQVEELQ KSLQDQGSKA EDSVLLKKKL EEHLEKLHEA NNELQKKRAI IEDLEPRFNN SSLKIEELQE ALRKKEEEMK QMEERYKKYL EKAKSVIRTL DPKQNQGAAP EIQALKNQLQ ERDRLFHSLE KEYEKTKSQR EMEEKYIVSA WYNMGMTLHK KAAEDRLAST GSGQSFLARQ RQATSSRRSY PGHVQPATAR
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: h-hook3; hHK3; hook homolog 3; mHK3; Protein Hook homolog 3
Gene Aliases: 5830454D03Rik; AI317159; E330005F07Rik; HK3; HOOK3
UniProt ID: (Human) Q86VS8, (Mouse) Q8BUK6
Entrez Gene ID: (Human) 84376, (Rat) 306548, (Mouse) 320191
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support