Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: QNQGAAPEIQ ALKNQLQERD RLFHSLEKEY EKTKSQREME EKYIVSAWYN MGMTLHKKAA EDRLASTGSG QSFLARQRQA TSSRRSYPGH VQPATAR
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: h-hook3; hHK3; hook homolog 3; Protein Hook homolog 3
Gene Aliases: HK3; HOOK3
UniProt ID: (Human) Q86VS8
Entrez Gene ID: (Human) 84376
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support