Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: EYPILSSGDS GTCSARAYPS DHRITTFQSC AVSANSCGGD DRFLVGRGVQ IGSPHHHHHH HHHHPQPATY QTSGNLGVSY SHSSCGPSYG SQNFSAPYSP YALNQEADV
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development. Two transcript variants encoding two different isoforms have been found for this gene, with only one of the isoforms containing the homeodomain region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box A1; homeobox 1F; Homeobox protein Hox-1F; Homeobox protein Hox-A1; Hox 1.6-like protein; HOX A1 homeodomain protein; HOXA 1; HOXA-1; lab-like protein; MGC45232
Gene Aliases: BSAS; HOX1; HOX1F; HOXA1
UniProt ID: (Human) P49639
Entrez Gene ID: (Human) 3198
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support