Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
Antibody detects endogenous levels of total HOXB3.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box 2G; homeo box B3; Homeobox protein Hox-2.7; Homeobox protein Hox-2G; Homeobox protein Hox-B3; Homeobox protein MH-23
Gene Aliases: Hox-2.7; HOX2; HOX2G; Hoxb-3; HOXB3
UniProt ID: (Human) P14651, (Mouse) P09026
Entrez Gene ID: (Human) 3213, (Mouse) 15410, (Rat) 303488
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support