Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The homeobox genes encode transcription factors that regulate developmental processes such as body patterning and organogenesis. The mammalian HOX gene complex consists of 39 genes that are located on 4 linkage groups (clusters), dispersed over 4 chromosomes (HOXA, HOXB, HOXC and HOXD). HOXB7 is expressed in most malignant T-cell lines and CD4-positive peripheral blood cells, and is constitutively expressed in both melanoma primary lesions and cell lines. HOXB7 binds to and transactivates bFGF in melanoma cell lines but SkBr3 breast adenocarcinoma is negative for the expression of both HOXB7 and bFGF genes. HOXB7 may be a key activator of tumor-associated neoangiogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box 2C; homeo box B7; homeo box c1 protein; Homeobox protein HHO.C1; Homeobox protein Hox-2.3; Homeobox protein Hox-2C; Homeobox protein Hox-B7; Homeobox protein MH-22B; Homeobox protein MuB1; Hox-23
Gene Aliases: AI325018; HHO.C1; Hox-2.3; HOX2; HOX2C; Hoxb-7; HOXB7
UniProt ID: (Human) P09629, (Mouse) P09024
Entrez Gene ID: (Human) 3217, (Mouse) 15415
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support