Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm's tumor and congenital vertical talus and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: d10; homeo box 4D; homeo box D10; Homeobox protein Hox-4.5; Homeobox protein Hox-4D; Homeobox protein Hox-4E; Homeobox protein Hox-5.3; Homeobox protein Hox-D10; Hox-45
Gene Aliases: AI385591; AI874987; Hox-4.4; Hox-4.5; Hox-5.3; HOX4; HOX4D; HOX4E; Hoxd-10; HOXD10
UniProt ID: (Human) P28358, (Mouse) P28359
Entrez Gene ID: (Human) 3236, (Mouse) 15430
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support