Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
HOX13D antibody (18736-1-AP) is produced by immunizing animals the N-terminal region of HOX13D and detects a 36 kDa band in cell and tissues.
HOXD13 (homeobox protein D13) belongs to a class of transcription factors called homeobox proteins. In vertebrates, homeobox are found in clusters called A, B, C, and D on four separate chromosomes. Expression of homeobox proteins is spatially and temporally regulated during embryonic development. The gene encoding HOXD13 is part of the D cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. Mutations in the HOXD13 gene cause synpolydactyly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box 4I; homeo box D13; Homeobox protein Hox-4.8; Homeobox protein Hox-4I; Homeobox protein Hox-D13
Gene Aliases: BDE; BDSD; Hox-4.8; HOX4I; HOXD13; SPD; SPD1; spdh
UniProt ID: (Human) P35453, (Mouse) P70217
Entrez Gene ID: (Human) 3239, (Rat) 288154, (Mouse) 15433
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support