Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Peptide Sequence: GKKQNKKKVE EVLEEEEEEY VVEKVLDRRV VKGKVEYLLK WKGFSDEDNT WEPEENLDCP DLIAEFLQSQ KTAHETDKSE GGKRKADSDS EDKGEESK
This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family. The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The protein may play an important role in the epigenetic control of chromatin structure and gene expression. Several related pseudogenes are located on chromosomes 1, 3, and X. Multiple alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromobox homolog 1; chromobox homolog 1 (HP1 beta homolog Drosophila ); Chromobox protein homolog 1; Heterochromatin protein 1 homolog beta; heterochromatin protein 1-beta; Heterochromatin protein p25; heterochromatin protein p25 beta; HGNC:1551; HP1 beta; HP1 beta homolog; HP1Hsbeta; M31; Modifier 1 protein; p25beta
Gene Aliases: CBX; CBX1; HP1-BETA; HP1Hs-beta; HP1Hsbeta; M31; MOD1; p25beta
UniProt ID: (Human) P83916
Entrez Gene ID: (Human) 10951, (Rat) 360609
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support