Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
HP1BP3 is the component of heterochromatin, may be involved in chromatin structure and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heterochromatin protein 1, binding protein 3; Heterochromatin protein 1-binding protein 3; HP1-BP74; Protein HP1-BP74
Gene Aliases: HP1-BP74; HP1BP3; HP1BP74
UniProt ID: (Human) Q5SSJ5
Entrez Gene ID: (Human) 50809
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support