Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: LILLGSPKLR TLARGLSPAY LRFGGTKTDF LIFDPKKEST FEERSYWQSQ VNQDICKYGS IPPDVEEKLR LEWPYQEQLL LREHYQKKFK NSTYSRSSV
Stem cells offer a therapeutic promise for many diseases. In the case of diabetes, stem cells offer the promise that beta cells may be generated ex vivo for the possible transplantation and cure of Type I diabetes (Couri CE, et al. ). For this to happen, stem cells will need to be isolated and expanded, they will need to be differentiated toward the beta cell lineage, and the differentiated cell progeny used for transplantation may need to be isolated from complex cell mixtures following differentiation. Appropriate markers targeting cell surface molecules on developing and mature pancreatic cells will be required for such research.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endo-glucoronidase; Heparanase; Heparanase-1; Hpa1
Gene Aliases: HEP; HPA; HPA1; HPR1; HPSE; HPSE1; HSE1
UniProt ID: (Human) Q9Y251
Entrez Gene ID: (Human) 10855
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support