Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: AAAEGSSRDY VAVNRSHGQL KTDEQGLNTH GASEISEDQM LGDSQSFSRT DSDTTTETAP GKGKLRSGSN TGLDYIKFTW KRLRSHSRQY VSGLHMNRER KAA
Histamine is a ubiquitous messenger molecule released from mast cells, enterochromaffin-like cells, and neurons. Its various actions are mediated by histamine receptors H1, H2, H3 and H4. This gene was thought to be intronless until recently. The protein encoded by this gene is an integral membrane protein and belongs to the G protein-coupled receptor superfamily. It mediates the contraction of smooth muscles, the increase in capillary permeability due to contraction of terminal venules, the release of catecholamine from adrenal medulla, and neurotransmission in the central nervous system. Multiple alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H1 receptor; H1-R; Histamine H1 receptor; Histamine H1-R; histamine receptor; histamine receptor, subclass H1
Gene Aliases: H1-R; H1R; HH1R; hisH1; HRH1
UniProt ID: (Human) P35367
Entrez Gene ID: (Human) 3269
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support