Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: filaggrin family member 3; Hornerin; intermediate filament-associated protein
Gene Aliases: FLG3; HRNR; S100A16; S100A18
UniProt ID: (Human) Q86YZ3
Entrez Gene ID: (Human) 388697
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support