Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: WRTRTTILQ STGKNFSKNI FAILQSVKAR EEGRAPEQRP APNAAPVDPT LRTKQPIPAA YNRYDQERFK GKEETEGFKI DTMGTYHGMT LKSVTEGASA RKTQTPAAQP VPRPVSQARP PPNQKKGSRT PIIIIPAATT SLITMLNAKD LLQDLKFVPS DEKKKQGCQR ENETLIQRRK DQMQPGGTAI SVTVPYRVVD QPLKLMPQDW DRVVAVFVQG PAWQFKGWPW LLPDGSPVDI FAKIKAFHLK YDEVRLDPNV QKWDVTVLEL SYHKRHLDRP VFLRFWETLD RYMVKHKSHL RF (232-531 aa encoded by BC014351)
Cdc73 is a tumor suppressor probably involved in transcriptional and post-transcriptional control pathways. Cdc73 is part of the PAF1 complex, a five-subunit protein complex composed of Paf1, Cdc73, Leo1, Rtf1 and Ctr9. PAF1 was first purified from yeast in association with RNA polymerase II and is believed to function in transcription elongation. The yeast PAF (yPAF) complex interacts with RNA polymerase II and coordinates the setting of histone marks associated with active transcription. In addition to coordinating events during transcription (initiation, promoter clearance, and elongation), hPAF also coordinates events in RNA quality control. Cdc73 may also be involved in cell cycle progression through the regulation of cyclin D1/PRAD1 expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell division cycle 73 Paf1/RNA polymerase II complex component-like protein; cell division cycle 73, Paf1/RNA polymerase II complex component, homolog; Cell division cycle protein 73 homolog; hyperparathyroidism 2 (with jaw tumor); hyperparathyroidism 2 homolog; Hyperparathyroidism 2 protein; Hyperparathyroidism 2 protein homolog; Paf1/RNA polymerase II complex component; Parafibromin; Vcell division cycle 73, Paf1/RNA polymerase II complex component, homolog
Gene Aliases: 8430414L16Rik; BC027756; C130030P16Rik; C1orf28; C81219; CDC73; FIHP; HPTJT; HRPT1; HRPT2; HYX
UniProt ID: (Human) Q6P1J9, (Mouse) Q8JZM7, (Rat) Q4V8C8
Entrez Gene ID: (Human) 79577, (Mouse) 214498, (Rat) 304832
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support