Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GCDFTNVVKT TVLLADINDF NTVNEIYKQY FKSNFPARAA YQVAALPKGS RIEIEAVAIQ GP
Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 89%.
Endoribonuclease responsible for the inhibition of the translation by cleaving mRNA. Inhibits cell-free protein synthesis. Cleaves phosphodiester bonds only in single-stranded RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 14.5 kDa translational inhibitor protein; 2-iminobutanoate/2-iminopropanoate deaminase; Heat-responsive protein 12; hp14.5; L-PSP; Liver perchloric acid-soluble protein; perchloric acid soluble protein; perchloric acid-soluble protein; perchrolic acid soluble protein; Reactive intermediate imine deaminase A homolog; reactive intermediate/imine deaminase A homolog; ribonuclease UK114; rp14.5; Translation inhibitor L-PSP ribonuclease; translational inhibitor p14.5; translational inhibitor protein p14.5; UK114 antigen homolog
Gene Aliases: HR12; HRP12; HRSP12; P14.5; PSP; PSP1; RIDA; UK114
UniProt ID: (Mouse) P52760, (Rat) P52759, (Human) P52758
Entrez Gene ID: (Mouse) 15473, (Rat) 65151, (Human) 10247
Molecular Function:
deaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support