Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MEETMKLATM EDTVEYCLFL IPDESRDSDK HKEILQKYIE RIITRFAPML VPYIWQNQPF NLKYKPGKGG VPAHMFGVTK FGDNIEDEWF IVYVIKQITK EFPELVARIE DNDGEFLLIE AADFLPKWLD PENSTNRVFF CHGELCIIPA PRKSGAESWL PTTPPTIPQA LNIITAHSEK ILASESIRAA VNRRIRGYPE KIQASLHRAH CFLPAGIVAV LKQRPRLVAA AVQAFYLRDP IDLRACRVFK TFLPETRIMT SVTFTKCLYA QLVQQRFVPD RRSGYRLPPP SDPQYRAHEL GMKLAHGFEI LCSKCSPHFS DCKKSLVTAS PLWASFLESL KKNDYFKGLI EGSAQYRERL EMAENYFQLS VDWPESSLAM SPGEEILTLL QTIPFDIEDL KKEAANLPPE DDDQWLDLSP DQLDQLLQEA VGKKESESVS KEEKEQNYDL TEVSESMKAF ISKVSTHKGA ELPREPSEAP ITFDADSFLN YFDKILGPRP NESDSDDLDD EDFECLDSDD DLDFETHEPG EEASLKGTLD NLKSYMAQMD QELAHTCISK SFTTRNQVEP VSQTTDNNSD EEDSGTGESV MAPVDVDLNL VSNILESYSS QAGLAGPASN LLQSMGVQLP DNTDHRPTSK PTKN
Regulator of p53/TP53 stability and function. Inhibits MDM2-mediated degradation of p53/TP53 possibly by cooperating in part with TXNIP (PubMed:16849563, PubMed:23880345). May be involved transcriptional regulation. In vitro has intrinsic transactivation activity enhanced by EP300. May be a transcriptional activator required for the expression of glycolytic genes (PubMed:19919181, PubMed:9928932). Involved in regulation of cell cycle progression. Proposed to disrupt Rb-E2F binding leading to transcriptional activation of E2F proteins (PubMed:19640839). The cell cycle -regulating function may depend on its RUVBL1-mediated association with the R2TP complex (PubMed:26711270). May play a role in regulation of pre-mRNA splicing (PubMed:24722212). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ecdysoneless homolog; hSGT1; Human suppressor of GCR two; Protein ecdysoneless homolog; protein SGT1; SGT1 protein; suppressor of GCR2; suppressor of S. cerevisiae gcr2
Gene Aliases: ECD; GCR2; HSGT1; SGT1
UniProt ID: (Human) O95905
Entrez Gene ID: (Human) 11319
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support