Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
A 1:5000 dilution of MA5-45122 was sufficient for detection of HSP27 in 20 µg of HeLa cell lysate by ECL immunoblot analysis.|Detects approximately 25kDa or approximately 27kDa. Recognizes HSP25 and HSP27, cross reacts with alpha B crystallin.
In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock or stress proteins. HSPB1 (heat shock protein beta-1 or HSP27) is a small heat shock protein which functions as a molecular chaperone that maintains denatured proteins in a folding-competent state. It plays a role in stress resistance and actin organization. Through its molecular chaperone activity, HSP27 regulates numerous biological processes including the phosphorylation and the axonal transport of neurofilament proteins. Mutations in the gene can result in Charcot-Marie-Tooth disease 2F and Neuronopathy distal hereditary motor 2B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 27 kDa heat shock protein 1; 28 kDa heat shock protein; DKFZp586P1322; epididymis secretory protein Li 102; Estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; Heat shock 25 kDa protein; Heat shock 27 kDa protein; heat shock 27 kDa protein 1; heat shock 27kD protein 1; heat shock 27kDa protein 1; Heat shock protein; heat shock protein 27 kDa beta-1; Heat shock protein beta-1; heat shock protein family B member 1 variant 1; heat shock protein family B member 1 variant 2; heat shock protein, 25 kDa; HS; HSP; HSP 25; HSP 27; HspB1; p25; Phospho-HSP 27; SRP27; Stress-responsive protein 27; truncated hsp25
Gene Aliases: 27kDa; CMT2F; HEL-S-102; HMN2B; HS.76067; Hsp25; HSP27; HSP28; HSPB1; I79_007264; SRP27
UniProt ID: (Human) P04792, (Mouse) P14602
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support