Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPVANAVIS VPAEFDLKQR NSTIEAANLA GLKILRVINE PTAAAMAYGL HKADVFHVLV IDLGGGTLDV SLLNKQGGMF LTRAMSGNNK LGGQDFNQRL LQYLYKQIYQ TYGFVPSRKE EIHRLRQAVE MVKLNLTLHQ SAQLSVLLTV EEQDRKEPHS SDTELPKDKL SSADDHRVNS GFGRGLSDKK SGESQVLFET EISRKLFDTL NEDLFQKILV PIQQVLKEGH LEKTEIDEVV LVGGSTRIPR IRQVIQEFFG KDPNTSVDPD LAVVTGVAIQ AGIDGGFWPL QVSALEIPNK HLQKTNFN (165-471 aa encoded by BC036370)
STCH encodes a protein that is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock 70 kDa protein 13; heat shock protein 13; heat shock protein 70kDa family member 13; heat shock protein 70kDa family, member 13; microsomal stress 70 protein ATPase core; Microsomal stress-70 protein ATPase core; stress 70 protein chaperone microsome-associated 60 kDa protein; stress 70 protein chaperone microsome-associated 60kD human homolog; stress 70 protein chaperone, microsome-associated, 60kD; stress 70 protein chaperone, microsome-associated, 60kD human homolog; stress 70 protein chaperone, microsome-associated, 60kDa; stress 70 protein chaperone, microsome-associated, human homolog; Stress-70 protein chaperone microsome-associated 60 kDa protein; testis secretory sperm-binding protein Li 199a
Gene Aliases: 1600002I10Rik; AV006182; B230217N24Rik; HSPA13; STCH
UniProt ID: (Human) P48723, (Rat) O35162, (Mouse) Q8BM72
Entrez Gene ID: (Human) 6782, (Rat) 29734, (Mouse) 110920
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support