Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody well recognizes the endogenous HSPA1L protein in multiple cell lines.
Immunogen sequence: STRIPKVQR LLQDYFNGRD LNKSINPDEA VAYGAAVQAA ILMGDKSEKV QDLLLLDVAP LSLGLETAGG VMTALMKRNS TIPTKQTQIF TTYSDNQPGV LIQVYEGERA MTKDNNLLGR FDLTGIPPAP RGVPQIEVTF DIDANGILNV TAMDKSTGKV NKITIANDKG RLSKEEIERM VLDAEKYKAE DEVQREKIAA KNALESYAFN MKSVVSDEGL KGKISESDKN KILDKCNELL SWLEVNQLAE KDEFDHKRKE LEQMCNPIIT KLYQGGCTGP ACGTGYVPGR PATGPTIEEV D (342-641 aa encoded by BC034483)
In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock proteins. One class of stress proteins, termed the "small heat shock proteins" is comprised of a diverse group of proteins from approximately15 to >30 kDa, with a single form found in yeast and human cells and multiple related forms found in higher plants. A direct relationship between elevated strwess protein expression and neoplasia has been suggested from studies on estrogen-induced proteins in breast cancer cells. In addition, Hsp27 exhibits rapid increased phosphorylation in response to various mitogens, tumor promoter (e. g. phorbol esters) and calcium ionophores. The expression of a 24 kDa protein was shown to correlate with steroid hormone receptor in cell lines, organs, and tumors. The 24 kDa estrogen-induced protein was found to be expressed at high levels in carcinoma of the breast. Another study has suggested that levels of the Hsp27 correlate with endometrial adenocarcinomas. Interestingly, the regulation of human Hsp27 by estrogens and heat shock is reminiscent of the ecdysteroid and heat shock activation of the drosophila Hsp20 genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heat shock 10kDa protein 1-like; Heat shock 70 kDa protein 1-Hom; Heat shock 70 kDa protein 1-like; Heat shock 70 kDa protein 1L; Heat shock 70 kDa protein 3; Heat shock 70 kDa-like protein 1; heat shock 70kD protein-like 1; heat shock 70kDa protein 1-like; Heat shock protein; Heat shock protein 70-1l; heat shock protein cognate 70, testis; heat shock-related protein hsc70t; HSP70-Hom; HSP70.3; inducible heat shock protein 70; Spermatid-specific heat shock protein 70
Gene Aliases: Hsc70t; HSP70-1L; Hsp70-3; HSP70-HOM; HSP70T; HSPA1L; hum70t; Msh5
UniProt ID: (Human) P34931, (Rat) P55063, (Mouse) P16627
Entrez Gene ID: (Human) 3305, (Rat) 24963, (Mouse) 15482
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support