Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 5
Peptide Sequence: MSVVGIDLGF QSCYVAVARA GGIETIANEY SDRCTPACIS FGPKNRSIGA AAKSQVISNA KNTVQGFKRF HGRAFSDPFV EAEKSNLAYD IVQLPTGLTG IKVTYMEEEQ NGPVDGQGDN PGPQAAEQGT DTAVPSDSDK KLPEMDID
The HSP 70 family is composed of four highly conserved proteins: HSP 70, HSC 70, GRP 75 and GRP 78. HSP 70 expression is strongly induced in response to heat stress. HSP 70 and HSC 70 play key roles in the cytosolic endoplasmic reticulum and mitochondrial import machinery and are found in both the cytosol and nucleus of mammalian cells. GRP 78 is localized in the endoplasmic reticulum, where it receives imported secretory proteins and is involved in the folding and translocation of nascent peptide chains. GRP 75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. It has been postulated that members of the HSP 70 family act as force-generating motors, relying on the hydrolysis of ATP for their activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein Li 5a; Heat shock 70 kDa protein 4; Heat shock 70-related protein APG-2; heat shock 70kD protein 4; heat shock 70kDa protein 4; Heat shock protein; Heat shock protein family H member 2; heat shock protein, 110 kDa; hsp70 RY; HSP70RY
Gene Aliases: APG-2; APG2; HEL-S-5a; HS24/P52; hsp70; hsp70RY; HSPA4; HSPH2; RY
UniProt ID: (Human) P34932
Entrez Gene ID: (Human) 3308
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support