Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The HSP70 family is composed of four highly conserved proteins: HSP70, HSC70, GRP75 and GRP78. These proteins serve a variety of roles. GRP75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. GRP75 and GRP78 are unresponsive to heat stress and are induced by glucose deprivation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 75 kDa glucose-regulated protein; catecholamine-regulated protein 40; epididymis secretory sperm binding protein Li 124m; GRP 75; GRP-75; Heat shock 70 kDa protein 9; heat shock 70kD protein 9B; heat shock 70kDa protein 9 (mortalin); heat shock 70kDa protein 9A; Heat shock protein; heat shock protein, A; HSP; MGC4500; Mortalin; mortalin, perinuclear; mortalin-2; mortalin2; MOT; mtHSP70; p66-mortalin; PBP74; Peptide-binding protein 74; Stress-70 protein, mitochondrial
Gene Aliases: CRP40; CSA; EVPLS; GRP-75; GRP75; HEL-S-124m; HSPA9; Hspa9a; HSPA9B; MOT; MOT2; mt-HSP70; MTHSP75; PBP74; SAAN; SIDBA4
UniProt ID: (Human) P38646
Entrez Gene ID: (Dog) 474697, (Human) 3313, (Rat) 291671
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support