Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
HSPB11 (Heat shock protein beta-11), also known as IFT25, was originally identified as a small heat shock protein. Recently it has been identified as a component of IFT complex B. It has been demonstrated that IFT25 is not required for ciliary assembly but is required for proper Hedgehog signaling, which in mammals occurs within cilia. Cilia lacking IFT25 have defects in the signal-dependent transport of multiple Hedgehog components and fail to activate the pathway upon stimulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock protein; Heat shock protein beta-11; heat shock protein family B (small), member 11; Heat shock protein family B member 11; HSP; Hspb11; intraflagellar transport 25 homolog; Intraflagellar transport protein 25 homolog; Placental protein 25; PP25
Gene Aliases: C1orf41; HSPB11; HSPC034; HSPCO34; IFT25; PP25
UniProt ID: (Human) Q9Y547
Entrez Gene ID: (Human) 51668
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support