Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DTVHIPEAT KQVVKAFQEF IRSTPYSAYD PETYTGHWKQ LTVRTSRRHQ AMAIAYFHPQ KLSPEELAEL KTSLAQHFTA GPGRASGVTC LYFVEEGQRK TPSQEGLPLE HVAGDRCIHE DLLGLTFRIS PHAFFQVNTP AAEVLYTVIQ DWAQLDAGSM VLDVCCGTGT IGLALARKVK RVIGVELCPE AVEDARVNAQ DNELSNVEFH CGRAEDLVPT LVSRLASQHL VAILDPPRAG LHSKVILAIR RAKNLRRLLY VSCNPRAAMG NFVDLCRAPS NRVKGIPFRP VKAVAVDLFP QTPHCEMLIL FERVEHPNGT GVLGPHSPPA QPTPGPPDNT LQETGTFPSS (277-625 aa encoded by BC013352)
The protein encoded by TRMT2A is of unknown function. However, it is orthologous to the mouse Trmt2a gene and contains an RNA methyltransferase domain. Expression of TRMT2A varies during the cell cycle, with aberrant expression being a possible biomarker in certain breast cancers. Several transcript variants encoding two different isoforms have been found for TRMT2A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HpaII tiny fragments locus 9c; HpaII tiny fragments locus 9c protein; mRNA (uracil-5-)-methyltransferase TRMT2A; TRM2 tRNA methyltransferase 2 homolog A; tRNA (uracil-5-)-methyltransferase homolog A
Gene Aliases: Htf9-c; HTF9C; TRMT2A
UniProt ID: (Human) Q8IZ69, (Mouse) Q8BNV1
Entrez Gene ID: (Human) 27037, (Rat) 287953, (Mouse) 15547
Molecular Function:
RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support