Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GYINSGLNP FLYAFLNKSF RRAFLIILCC DDERYRRPSI LGQTVPCSTT TINGSTHVLR DAVECGGQWE SQCHPPATSP LVAAQPSDT (301-388 aa encoded by BC074755)
This gene is a member of the family of serotonin receptors, which are G protein coupled receptors that stimulate cAMP production in response to serotonin (5-hydroxytryptamine). The gene product is a glycosylated transmembrane protein that functions in both the peripheral and central nervous system to modulate the release of various neurotransmitters. Multiple transcript variants encoding proteins with distinct C-terminal sequences have been described, but the full-length nature of some transcript variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-HT-4; 5-HT4 receptor; 5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled; 5-hydroxytryptamine receptor 4; 5HT4; 5HT4 receptor; cardiac 5-HT4 receptor; Serotonin receptor 4
Gene Aliases: 5-HT-4; 5-HT4; 5-HT4R; 5-HT<4L>; 5HTR4; HTR4
UniProt ID: (Human) Q13639, (Mouse) P97288
Entrez Gene ID: (Human) 3360, (Mouse) 15562
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support