Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AAVPSPPPA SPRSQYNFIA DVVEKTAPAV VYIEILDRHP FLGREVPISN GSGFVVAADG LIVTNAHVVA DRRRVRVRLL SGDTYEAVVT AVDPVADIAT LRIQTKEPLP TLPLGRSADV RQGEFVVAMG SPFALQNTIT SGIVSSAQRP ARDLGLPQTN VEYIQTDAAI DFGNSGGPLV NLDGEVIGVN TMKVTAGISF AIPSDRLREF LHRGEKKNSS SGISGSQRRY IGVMMLTLSP SILAELQLRE PSFPDVQHGV LIHKVILGSP AHRAGLRPGD VILAIGEQMV QNAEDVYEAV RTQSQLAVQI RRGRETLTLY VTPEVTE (133-458 aa encoded by BC000096 )
The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity (1-3). Recently, a serine protease has been isolated, which is released from the mitochondria upon induction of apoptosis by apoptotic stimuli. This protein called Omi/HtrA2 is a mitochondrial protein that binds to IAP (4). Like Smac/DIABLO, the mature Omi protein contains a conserved IAP-binding motif (AVPS) at its N terminus. The deregulated expression of Omi in the cytoplasm of mammalian cells induces apoptosis indicating that Omi could participate in the mitochondrial apoptotic pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High temperature requirement protein A2; HtrA-like serine protease; HtrA2; motor neuron degeneration 2; Omi stress-regulated endoprotease; protease, serine, 25; Serine protease 25; Serine protease HTRA2, mitochondrial; serine protease OMI; Serine proteinase OMI
Gene Aliases: AI481710; HTRA2; mnd2; OMI; PARK13; PRSS25
UniProt ID: (Human) O43464, (Mouse) Q9JIY5
Entrez Gene ID: (Human) 27429, (Rat) 297376, (Mouse) 64704
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support