Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTSRRLEES MGAVQMGLVN MFKGFQSKVL PPLSPKVVTE EEVNRMLTPS EFLKEMSLTT EQRLAKTRLM CRPQIIELLD MGETTHQKFS GIDLDQALFQ PFPSEIIFQN YTPCEVYEVP LILRNNDKIP RLVKVVEESS PYFKVISPKD IGHKVAPGVP SIFRILFTPE ENKDYAHTLT CVTEREKFIV PIKARGARAI LDFPDKLNFS TCPVKYSTQK ILLVRNIGNK NAVFHIKTCR PFSIEPAIGT LNVGESMQLE VEFEPQSVGD HSGRLIVCYD TGEKVFVSLY GAAIDMNIRL DKNSLTIEKT YISMANQRTI TIHNRSNIIA HFLWKVFATQ QEEDRE (22-366 aa encoded by BC028351)
Required for ciliary motility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hydrocephalus-inducing protein homolog; protein phosphatase 1, regulatory subunit 31
Gene Aliases: CILD5; HYDIN; HYDIN1; HYDIN2; KIAA1864; PPP1R31
UniProt ID: (Human) Q4G0P3
Entrez Gene ID: (Human) 54768
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support