Invitrogen
The MA1-7456 antibody reacts with Helicobacter pylori 19 kDa Ag in bacterial and infected tissue samples.
MA1-7456 has been successfully used in ELISA, Immunofluorescence and Western blot applications.
The MA1-7456 antibody was raised against H. pylori 19 kDa antigen.
Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
Protein Aliases: H. pylori