Invitrogen
FIGURE: 1 / 3
The PA1-73161 antibody reacts with Helicobacter pylori.
PA1-73161 has been successfully used in Immunofluorescence applications.
The PA1-73161 antibody was raised against whole H. pylori cells; ATCC strain 43504.
Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
Protein Aliases: H. pylori