Invitrogen
The PA1-73162 antibody reacts with Helicobacter pylori.
PA1-73162 has been successfully used in ELISA applications.
The PA1-73162 antibody was raised against whole H. pylori cells; ATCC strain 43504.
Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Protein Aliases: H. pylori