Search Thermo Fisher Scientific
The PA1-73107 antibody reacts with Hepatitis C virus NS3 antigen.
PA1-73107 has been successfully used in ELISA, immunofluorescence, Western blot and Immunohistochemical applications.
The PA1-73107 antibody was raised against recombinant NS3 (genotype 1a).
Hepatitis C virus (HCV) is a small (55-65 nm in size), enveloped, positive-sense single-stranded RNA virus of the family Flaviviridae. The hepatitis C virus is the cause of hepatitis C and some cancers such as liver cancer (hepatocellular carcinoma, abbreviated HCC) and lymphomas in humans. Nonstructural protein 3 (NS3), also known as p-70, is a viral nonstructural protein that is 70 kDa cleavage product of the hepatitis C virus polyprotein. It acts as a serine protease. C-terminal two thirds of the protein also acts as helicase and nucleoside triphosphatase. First (N-terminal) 180 aminoacids of NS3 has additional role as cofactor domains for NS2 protein.
Protein Aliases: HCV; HCV NS3; Hepatitis C Virus; HepC; HepC NS3