Bioss
Nonstructural protein 5B is a viral protein found in the hepatitis C virus. It is an RNA-dependent RNA polymerase, having the key function of replicating HCV's viral RNA by using the viral positive RNA strand as a template to catalyze the polymerization of ribonucleoside triphosphates during RNA replication.