Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TYQWNIPER SGPGPNDSAC VSWIYYSAVD PIKDMYSGLV GPLAICQKGI LEPHGGRSDM DREFALLFLI FDENKSWYLE ENVATHGSQD PGSINLQDET FLESNKMHAI NGKLYANLRG LTMYQGERVA WYMLAMGQDV DLHTIHFHAE SFLYRNGENY RADVVDLFPG TFEVVEMVAS NPGTWLMHCH VTDHVHAGME TLFTVFSRTE HLSPLTVITK ETEKAVPPRD IEEGNVKMLG MQIPIKNVEM LASVLVAISV TLLLVVLALG GVVWYQHRQR KLRRNRRSIL DDSFKLLSFK Q (859-1158 aa encoded by BC011561)
B1AJX8(HEPH) is similar to an iron transport protein found in mouse. The mouse protein is similar to ceruloplasmin, a serum multi-copper ferroxidase, and is thought to be a membrane-bound protein responsible for transport of dietary iron from epithelial cells of the intestinal lumen into the circulatory system. In mouse, defects in this gene can lead to severe microcytic anemia. Three transcript variants encoding different isoforms have been described for this gene. The protein encoded by this gene is similar to an iron transport protein found in mouse. The mouse protein is similar to ceruloplasmin, a serum multi-copper ferroxidase, and is thought to be a membrane-bound protein responsible for transport of dietary iron from epithelial cells of the intestinal lumen into the circulatory system. In mouse, defects in this gene can lead to severe microcytic anemia. Two transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hephaestin; Hp
Gene Aliases: CPL; HEPH; KIAA0698; UNQ2562/PRO6242
UniProt ID: (Human) Q9BQS7
Entrez Gene ID: (Human) 9843
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support